Material Safety Data Sheet
L-Anabasine, 90%

ACC# 02161
Section 1 - Chemical Product and Company Identification
MSDS Name: L-Anabasine, 90%
Catalog Numbers: AC342601000, AC342605000
Synonyms:
Neonicotine2-(3-Pyridyl)piperidine
Company Identification:

              Acros Organics N.V.
              One Reagent Lane
              Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300

Section 2 - Composition, Information on Ingredients

CAS# Chemical Name Percent EINECS/ELINCS
40774-73-0 L-Anabasine 90% unlisted

Section 3 - Hazards Identification

EMERGENCY OVERVIEW

Appearance: light yellow liquid. Flash Point: 93 deg C.
Caution! Combustible liquid and vapor. May cause eye and skin irritation. May cause respiratory and digestive tract irritation. May cause central nervous system depression. Light sensitive. Hygroscopic (absorbs moisture from the air). The toxicological properties of this material have not been fully investigated.
Target Organs: Central nervous system.


Potential Health Effects
Eye: May cause eye irritation. May cause chemical conjunctivitis and corneal damage.
Skin: May cause skin irritation. Toxic in contact with skin. May cause cyanosis of the extremities.
Ingestion: Poison by ingestion. May cause gastrointestinal irritation with nausea, vomiting and diarrhea. The toxicological properties of this substance have not been fully investigated.
Inhalation: May cause respiratory tract irritation. The toxicological properties of this substance have not been fully investigated. Toxic if inhaled. Aspiration may lead to pulmonary edema. Inhalation at high concentrations may cause CNS depression and asphixiation.
Chronic: Effects may be delayed.

Section 4 - First Aid Measures


Eyes: Immediately flush eyes with plenty of water for at least 15 minutes, occasionally lifting the upper and lower eyelids. Get medical aid.
Skin: Get medical aid immediately. Immediately flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes.
Ingestion: Never give anything by mouth to an unconscious person. Get medical aid immediately. Do NOT induce vomiting. If conscious and alert, rinse mouth and drink 2-4 cupfuls of milk or water. Wash mouth out with water.
Inhalation: Get medical aid immediately. Remove from exposure and move to fresh air immediately. If breathing is difficult, give oxygen. Do NOT use mouth-to-mouth resuscitation. If breathing has ceased apply artificial respiration using oxygen and a suitable mechanical device such as a bag and a mask.
Notes to Physician: Treat symptomatically and supportively.

Section 5 - Fire Fighting Measures


General Information: As in any fire, wear a self-contained breathing apparatus in pressure-demand, MSHA/NIOSH (approved or equivalent), and full protective gear. During a fire, irritating and highly toxic gases may be generated by thermal decomposition or combustion. Use water spray to keep fire-exposed containers cool. Combustible liquid. Containers may explode when heated.
Extinguishing Media: Use water spray to cool fire-exposed containers. Use water spray, dry chemical, carbon dioxide, or chemical foam. Use agent most appropriate to extinguish fire.
Flash Point: 93 deg C ( 199.40 deg F)
Autoignition Temperature: Not available.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating: (estimated) Health: 2; Flammability: 2; Instability: 0

Section 6 - Accidental Release Measures


General Information: Use proper personal protective equipment as indicated in Section 8.
Spills/Leaks: Absorb spill with inert material (e.g. vermiculite, sand or earth), then place in suitable container. Avoid runoff into storm sewers and ditches which lead to waterways. Clean up spills immediately, observing precautions in the Protective Equipment section. Remove all sources of ignition. Use a spark-proof tool. Provide ventilation.

Section 7 - Handling and Storage


Handling: Wash thoroughly after handling. Do not breathe dust, vapor, mist, or gas. Do not get in eyes, on skin, or on clothing. Empty containers retain product residue, (liquid and/or vapor), and can be dangerous. Keep container tightly closed. Keep away from heat, sparks and flame. Avoid ingestion and inhalation. Store protected from light. Use only in a chemical fume hood. Do not pressurize, cut, weld, braze, solder, drill, grind, or expose empty containers to heat, sparks or open flames.
Storage: Keep away from sources of ignition. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Store protected from moisture. Store protected from light.

Section 8 - Exposure Controls, Personal Protection


Engineering Controls: Facilities storing or utilizing this material should be equipped with an eyewash facility and a safety shower. Use adequate ventilation to keep airborne concentrations low.
Exposure Limits
Chemical Name ACGIH NIOSH OSHA - Final PELs
L-Anabasine none listed none listed none listed

OSHA Vacated PELs: L-Anabasine: No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes: Wear appropriate protective eyeglasses or chemical safety goggles as described by OSHA's eye and face protection regulations in 29 CFR 1910.133 or European Standard EN166.
Skin: Wear appropriate protective gloves to prevent skin exposure.
Clothing: Wear appropriate protective clothing to prevent skin exposure.
Respirators: Follow the OSHA respirator regulations found in 29 CFR 1910.134 or European Standard EN 149. Use a NIOSH/MSHA or European Standard EN 149 approved respirator if exposure limits are exceeded or if irritation or other symptoms are experienced.

Section 9 - Physical and Chemical Properties


Physical State: Liquid
Appearance: light yellow
Odor: Not available.
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: Not available.
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: 270 deg C
Freezing/Melting Point:9 deg C
Decomposition Temperature:> 300 deg C
Solubility: Soluble.
Specific Gravity/Density:1.054
Molecular Formula:C10H14N2
Molecular Weight:162.23

Section 10 - Stability and Reactivity


Chemical Stability: Moisture sensitive. Light sensitive.
Conditions to Avoid: Incompatible materials, light, ignition sources, excess heat, exposure to moist air or water.
Incompatibilities with Other Materials: Moisture, oxidizing agents.
Hazardous Decomposition Products: Nitrogen oxides, carbon monoxide, irritating and toxic fumes and gases, carbon dioxide.
Hazardous Polymerization: Has not been reported

Section 11 - Toxicological Information


RTECS#:      
CAS# 40774-73-0 unlisted.
LD50/LC50:
Not available.

Carcinogenicity:
CAS# 40774-73-0: Not listed by ACGIH, IARC, NTP, or CA Prop 65.

Epidemiology: No information found
Teratogenicity: No information found
Reproductive Effects: No information found
Mutagenicity: No information found
Neurotoxicity: No information found
Other Studies:

Section 12 - Ecological Information

No information available.

Section 13 - Disposal Considerations

Chemical waste generators must determine whether a discarded chemical is classified as a hazardous waste. US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3. Additionally, waste generators must consult state and local hazardous waste regulations to ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.

Section 14 - Transport Information

US DOT Canada TDG
Shipping Name: DOT regulated - small quantity provisions apply (see 49CFR173.4) No information available.
Hazard Class:
UN Number:
Packing Group:

Section 15 - Regulatory Information

US FEDERAL

TSCA
     CAS# 40774-73-0 is not listed on the TSCA inventory. It is for research and development use only.
Health & Safety Reporting List
     None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
     None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
     None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
     None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
     None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
     None of the chemicals in this product have a TPQ.
Section 313      No chemicals are reportable under Section 313.
Clean Air Act:
     This material does not contain any hazardous air pollutants.
     This material does not contain any Class 1 Ozone depletors.
     This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
     None of the chemicals in this product are listed as Hazardous Substances under the CWA.
     None of the chemicals in this product are listed as Priority Pollutants under the CWA.
     None of the chemicals in this product are listed as Toxic Pollutants under the CWA.
OSHA:
     None of the chemicals in this product are considered highly hazardous by OSHA.
STATE
     CAS# 40774-73-0 is not present on state lists from CA, PA, MN, MA, FL, or NJ.

California Prop 65

California No Significant Risk Level: None of the chemicals in this product are listed.

European/International Regulations

European Labeling in Accordance with EC Directives
Hazard Symbols:
     T
Risk Phrases:

     R 23/24/25 Toxic by inhalation, in contact with skin and if
     swallowed.

Safety Phrases:

     S 36/37/39 Wear suitable protective clothing, gloves and eye/face pr
     otection.
     S 45 In case of accident or if you feel unwell, seek medical advice
     immediately (show the label where possible).
     S 28A After contact with skin, wash immediately with plenty of water
     .

WGK (Water Danger/Protection)

     CAS# 40774-73-0: No information available.
Canada - DSL/NDSL
None of the chemicals in this product are listed on the DSL or NDSL list.
Canada - WHMIS
     This product has a WHMIS classification of B3.
This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations and the MSDS contains all of the information required by those regulations.
Canadian Ingredient Disclosure List

Section 16 - Additional Information

MSDS Creation Date: 1/31/2000
Revision #6 Date: 3/04/2004

The information above is believed to be accurate and represents the best information currently available to us. However, we make no warranty of merchantability or any other warranty, express or implied, with respect to such information, and we assume no liability resulting from its use. Users should make their own investigations to determine the suitability of the information for their particular purposes. In no event shall Fisher be liable for any claims, losses, or damages of any third party or for lost profits or any special, indirect, incidental, consequential or exemplary damages, howsoever arising, even if Fisher has been advised of the possibility of such damages.