Material Safety Data Sheet
Palatinose

ACC# 39428
Section 1 - Chemical Product and Company Identification
MSDS Name: Palatinose
Catalog Numbers: AC225950000, AC225950050
Synonyms:
6-O-alpha-D-Glucopyranosyl-D-fructofuranose; Isomaltulose
Company Identification:

              Acros Organics N.V.
              One Reagent Lane
              Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300

Section 2 - Composition, Information on Ingredients

CAS# Chemical Name Percent EINECS/ELINCS
13718-94-0 Palatinose 237-282-1
58166-27-1 PALATINOSE 261-150-2

Section 3 - Hazards Identification

EMERGENCY OVERVIEW

Appearance: white to pale yellow solid.
Caution! May cause eye and skin irritation. May cause respiratory and digestive tract irritation. The toxicological properties of this material have not been fully investigated.
Target Organs: None.


Potential Health Effects
Eye: May cause eye irritation.
Skin: May cause skin irritation.
Ingestion: May cause irritation of the digestive tract. The toxicological properties of this substance have not been fully investigated.
Inhalation: May cause respiratory tract irritation.
Chronic: Not available.

Section 4 - First Aid Measures


Eyes: Flush eyes with plenty of water for at least 15 minutes, occasionally lifting the upper and lower eyelids. Get medical aid.
Skin: Flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes. Get medical aid if irritation develops or persists.
Ingestion: Get medical aid. Wash mouth out with water.
Inhalation: Remove from exposure and move to fresh air immediately. Get medical aid if cough or other symptoms appear.
Notes to Physician: Treat symptomatically and supportively.

Section 5 - Fire Fighting Measures


General Information: As in any fire, wear a self-contained breathing apparatus in pressure-demand, MSHA/NIOSH (approved or equivalent), and full protective gear.
Extinguishing Media: Use water spray, dry chemical, carbon dioxide, or chemical foam.
Flash Point: Not available.
Autoignition Temperature: Not available.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating: Not published.

Section 6 - Accidental Release Measures


General Information: Use proper personal protective equipment as indicated in Section 8.
Spills/Leaks: Vacuum or sweep up material and place into a suitable disposal container.

Section 7 - Handling and Storage


Handling: Avoid breathing dust, vapor, mist, or gas. Avoid contact with skin and eyes.
Storage: Store in a cool, dry place. Store in a tightly closed container.

Section 8 - Exposure Controls, Personal Protection


Engineering Controls: Facilities storing or utilizing this material should be equipped with an eyewash facility and a safety shower.
Exposure Limits
Chemical Name ACGIH NIOSH OSHA - Final PELs
Palatinose none listed none listed none listed
PALATINOSE none listed none listed none listed

OSHA Vacated PELs: Palatinose: No OSHA Vacated PELs are listed for this chemical. PALATINOSE: No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes: Wear chemical splash goggles.
Skin: Wear appropriate protective gloves to prevent skin exposure.
Clothing: Wear a chemical apron.
Respirators: A NIOSH/MSHA approved air purifying dust or mist respirator or European Standard EN 149.

Section 9 - Physical and Chemical Properties


Physical State: Solid
Appearance: white to pale yellow
Odor: Not available.
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: Not available.
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: Not available.
Freezing/Melting Point:125 - 128 deg C
Decomposition Temperature:Not available.
Solubility: Not available.
Specific Gravity/Density:Not available.
Molecular Formula:C12H22O11
Molecular Weight:342.29

Section 10 - Stability and Reactivity


Chemical Stability: Not available.
Conditions to Avoid: Not available.
Incompatibilities with Other Materials: Strong oxidizing agents.
Hazardous Decomposition Products: Carbon monoxide, carbon dioxide.
Hazardous Polymerization: Has not been reported.

Section 11 - Toxicological Information


RTECS#:      
CAS# 13718-94-0 unlisted.      
CAS# 58166-27-1 unlisted.
LD50/LC50:
Not available.
Not available.

Carcinogenicity:
CAS# 13718-94-0: Not listed by ACGIH, IARC, NTP, or CA Prop 65.
CAS# 58166-27-1: Not listed by ACGIH, IARC, NTP, or CA Prop 65.

Epidemiology: No information available.
Teratogenicity: No information available.
Reproductive Effects: No information available.
Mutagenicity: No information available.
Neurotoxicity: No information available.
Other Studies:

Section 12 - Ecological Information

No information available.

Section 13 - Disposal Considerations

Chemical waste generators must determine whether a discarded chemical is classified as a hazardous waste. US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3. Additionally, waste generators must consult state and local hazardous waste regulations to ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.

Section 14 - Transport Information

US DOT Canada TDG
Shipping Name: Not regulated as a hazardous material No information available.
Hazard Class:
UN Number:
Packing Group:

Section 15 - Regulatory Information

US FEDERAL

TSCA
     CAS# 13718-94-0 is not listed on the TSCA inventory. It is for research and development use only.
     CAS# 58166-27-1 is not listed on the TSCA inventory. It is for research and development use only.
Health & Safety Reporting List
     None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
     None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
     None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
     None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
     None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
     None of the chemicals in this product have a TPQ.
Section 313      No chemicals are reportable under Section 313.
Clean Air Act:
     This material does not contain any hazardous air pollutants.
     This material does not contain any Class 1 Ozone depletors.
     This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
     None of the chemicals in this product are listed as Hazardous Substances under the CWA.
     None of the chemicals in this product are listed as Priority Pollutants under the CWA.
     None of the chemicals in this product are listed as Toxic Pollutants under the CWA.
OSHA:
     None of the chemicals in this product are considered highly hazardous by OSHA.
STATE
     CAS# 13718-94-0 is not present on state lists from CA, PA, MN, MA, FL, or NJ.
     CAS# 58166-27-1 is not present on state lists from CA, PA, MN, MA, FL, or NJ.

California Prop 65

California No Significant Risk Level: None of the chemicals in this product are listed.

European/International Regulations

European Labeling in Accordance with EC Directives
Hazard Symbols:
      Not available.
Risk Phrases:


Safety Phrases:

     S 24/25 Avoid contact with skin and eyes.

WGK (Water Danger/Protection)

     CAS# 13718-94-0: No information available.
     CAS# 58166-27-1: No information available.
Canada - DSL/NDSL
None of the chemicals in this product are listed on the DSL or NDSL list.
Canada - WHMIS
     WHMIS: Not available.
This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations and the MSDS contains all of the information required by those regulations.
Canadian Ingredient Disclosure List

Section 16 - Additional Information

MSDS Creation Date: 9/02/1997
Revision #3 Date: 10/03/2005

The information above is believed to be accurate and represents the best information currently available to us. However, we make no warranty of merchantability or any other warranty, express or implied, with respect to such information, and we assume no liability resulting from its use. Users should make their own investigations to determine the suitability of the information for their particular purposes. In no event shall Fisher be liable for any claims, losses, or damages of any third party or for lost profits or any special, indirect, incidental, consequential or exemplary damages, howsoever arising, even if Fisher has been advised of the possibility of such damages.