Material Safety Data Sheet
Rhodium (II) acetate, dimer, anhydrous, ca. 43 % Rh

ACC# 95104
Section 1 - Chemical Product and Company Identification
MSDS Name: Rhodium (II) acetate, dimer, anhydrous, ca. 43 % Rh
Catalog Numbers: AC268630000, AC268630010, AC268632500, AC268635000
Synonyms:
Rhodium diacetate; RH-110; Rhodium(ii) acetate.
Company Identification:

              Acros Organics N.V.
              One Reagent Lane
              Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300

Section 2 - Composition, Information on Ingredients

CAS# Chemical Name Percent EINECS/ELINCS
15956-28-2 Rhodium (II) acetate, dimer, anhydrous, ca. 43 % Rh ca. 57.0240-084-8

Section 3 - Hazards Identification

EMERGENCY OVERVIEW

Appearance: green powder.
Warning! May cause burns by all exposure routes. Irritant. Causes eye and skin irritation. May cause respiratory and digestive tract irritation. May cause central nervous system effects. May cause cardiac disturbances. The toxicological properties of this material have not been fully investigated.
Target Organs: Central nervous system, cardiovascular system.


Potential Health Effects
Eye: Causes eye irritation and possible burns. May cause chemical conjunctivitis.
Skin: Causes skin irritation and possible burns.
Ingestion: May cause cardiac disturbances. The toxicological properties of this substance have not been fully investigated. May cause central nervous system effects. May cause irritation of the digestive tract and possible burns.
Inhalation: May cause respiratory tract irritation. The toxicological properties of this substance have not been fully investigated. Central nervous system effects, which appear to predominate in acute cases are characterized by abnormal fatigue, memory difficulties and dizziness.
Chronic: Effects may be delayed.

Section 4 - First Aid Measures


Eyes: Get medical aid. Do NOT allow victim to rub eyes or keep eyes closed. Extensive irrigation with water is required (at least 30 minutes).
Skin: Get medical aid. Immediately flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes. Wash clothing before reuse.
Ingestion: Never give anything by mouth to an unconscious person. Get medical aid. Do NOT induce vomiting. If conscious and alert, rinse mouth and drink 2-4 cupfuls of milk or water.
Inhalation: Remove from exposure and move to fresh air immediately. If breathing is difficult, give oxygen. Get medical aid. Do NOT use mouth-to-mouth resuscitation. If breathing has ceased apply artificial respiration using oxygen and a suitable mechanical device such as a bag and a mask.
Notes to Physician: Treat symptomatically and supportively.

Section 5 - Fire Fighting Measures


General Information: As in any fire, wear a self-contained breathing apparatus in pressure-demand, MSHA/NIOSH (approved or equivalent), and full protective gear. During a fire, irritating and highly toxic gases may be generated by thermal decomposition or combustion. Contact with metals may evolve flammable hydrogen gas. Containers may explode when heated. Non-combustible, substance itself does not burn but may decompose upon heating to produce irritating, corrosive and/or toxic fumes. Runoff from fire control or dilution water may cause pollution.
Extinguishing Media: Use water spray to cool fire-exposed containers. Do NOT get water inside containers. For small fires, use dry chemical, carbon dioxide, or water spray. For large fires, use dry chemical, carbon dioxide, alcohol-resistant foam, or water spray.
Flash Point: Not available.
Autoignition Temperature: Not available.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating: (estimated) Health: 3; Flammability: 0; Instability: 0

Section 6 - Accidental Release Measures


General Information: Use proper personal protective equipment as indicated in Section 8.
Spills/Leaks: Vacuum or sweep up material and place into a suitable disposal container. Clean up spills immediately, observing precautions in the Protective Equipment section. Avoid generating dusty conditions. Provide ventilation.

Section 7 - Handling and Storage


Handling: Wash thoroughly after handling. Minimize dust generation and accumulation. Avoid breathing dust, vapor, mist, or gas. Avoid contact with eyes, skin, and clothing. Keep container tightly closed. Avoid ingestion and inhalation. Use only in a chemical fume hood. Wash clothing before reuse.
Storage: Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Corrosives area.

Section 8 - Exposure Controls, Personal Protection


Engineering Controls: Facilities storing or utilizing this material should be equipped with an eyewash facility and a safety shower. Use adequate ventilation to keep airborne concentrations low. Use only under a chemical fume hood.
Exposure Limits
Chemical Name ACGIH NIOSH OSHA - Final PELs
Rhodium (II) acetate, dimer, anhydrous, ca. 43 % Rh none listed none listed none listed

OSHA Vacated PELs: Rhodium (II) acetate, dimer, anhydrous, ca. 43 % Rh: No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes: Wear appropriate protective eyeglasses or chemical safety goggles as described by OSHA's eye and face protection regulations in 29 CFR 1910.133 or European Standard EN166.
Skin: Wear appropriate protective gloves to prevent skin exposure.
Clothing: Wear a chemical apron. Wear appropriate protective clothing to prevent skin exposure.
Respirators: Follow the OSHA respirator regulations found in 29 CFR 1910.134 or European Standard EN 149. Use a NIOSH/MSHA or European Standard EN 149 approved respirator if exposure limits are exceeded or if irritation or other symptoms are experienced.

Section 9 - Physical and Chemical Properties


Physical State: Powder
Appearance: green
Odor: Not available.
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: 15.24
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: 88-90C
Freezing/Melting Point:Not available.
Decomposition Temperature:Not available.
Solubility: Not available.
Specific Gravity/Density:.888
Molecular Formula:(CH3Co2)Rh2
Molecular Weight:441.98

Section 10 - Stability and Reactivity


Chemical Stability: Stable at room temperature in closed containers under normal storage and handling conditions.
Conditions to Avoid: Incompatible materials, dust generation, excess heat, strong oxidants.
Incompatibilities with Other Materials: Oxidizing agents.
Hazardous Decomposition Products: Carbon monoxide, irritating and toxic fumes and gases, carbon dioxide, rhodium/rhodium oxides.
Hazardous Polymerization: Has not been reported

Section 11 - Toxicological Information


RTECS#:      
CAS# 15956-28-2: VI9361000
LD50/LC50:
Not available.

Carcinogenicity:
CAS# 15956-28-2: Not listed by ACGIH, IARC, NTP, or CA Prop 65.

Epidemiology: No information found
Teratogenicity: No information found
Reproductive Effects: No information found
Mutagenicity: No information found
Neurotoxicity: No information found
Other Studies:

Section 12 - Ecological Information

No information available.

Section 13 - Disposal Considerations

Chemical waste generators must determine whether a discarded chemical is classified as a hazardous waste. US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3. Additionally, waste generators must consult state and local hazardous waste regulations to ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.

Section 14 - Transport Information

US DOT Canada TDG
Shipping Name: DOT regulated - small quantity provisions apply (see 49CFR173.4) No information available.
Hazard Class:
UN Number:
Packing Group:

Section 15 - Regulatory Information

US FEDERAL

TSCA
     CAS# 15956-28-2 is not listed on the TSCA inventory. It is for research and development use only.
Health & Safety Reporting List
     None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
     None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
     None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
     None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
     None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
     None of the chemicals in this product have a TPQ.
Section 313      No chemicals are reportable under Section 313.
Clean Air Act:
     This material does not contain any hazardous air pollutants.
     This material does not contain any Class 1 Ozone depletors.
     This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
     None of the chemicals in this product are listed as Hazardous Substances under the CWA.
     None of the chemicals in this product are listed as Priority Pollutants under the CWA.
     None of the chemicals in this product are listed as Toxic Pollutants under the CWA.
OSHA:
     None of the chemicals in this product are considered highly hazardous by OSHA.
STATE
     CAS# 15956-28-2 is not present on state lists from CA, PA, MN, MA, FL, or NJ.

California Prop 65

California No Significant Risk Level: None of the chemicals in this product are listed.

European/International Regulations

European Labeling in Accordance with EC Directives
Hazard Symbols:
     XI
Risk Phrases:

     R 36/38 Irritating to eyes and skin.

Safety Phrases:

     S 15 Keep away from heat.
     S 26 In case of contact with eyes, rinse immediately with plenty of
     water and seek medical advice.
     S 37/39 Wear suitable gloves and eye/face protection.
     S 28A After contact with skin, wash immediately with plenty of water
     .

WGK (Water Danger/Protection)

     CAS# 15956-28-2: 0
Canada - DSL/NDSL
None of the chemicals in this product are listed on the DSL or NDSL list.
Canada - WHMIS
     WHMIS: Not available.
This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations and the MSDS contains all of the information required by those regulations.
Canadian Ingredient Disclosure List

Section 16 - Additional Information

MSDS Creation Date: 2/23/1999
Revision #4 Date: 10/03/2005

The information above is believed to be accurate and represents the best information currently available to us. However, we make no warranty of merchantability or any other warranty, express or implied, with respect to such information, and we assume no liability resulting from its use. Users should make their own investigations to determine the suitability of the information for their particular purposes. In no event shall Fisher be liable for any claims, losses, or damages of any third party or for lost profits or any special, indirect, incidental, consequential or exemplary damages, howsoever arising, even if Fisher has been advised of the possibility of such damages.