Material Safety Data Sheet
9(10H)-Acridone

ACC# 97301
Section 1 - Chemical Product and Company Identification
MSDS Name: 9(10H)-Acridone
Catalog Numbers: AC400250000, AC400250100, AC400250250
Synonyms:
9-Acridanone; Acridanone; 9(10H)-Acridanone (9CI); Acridone; 9-Acridone
Company Identification:

              Acros Organics N.V.
              One Reagent Lane
              Fair Lawn, NJ 07410
For information in North America, call: 800-ACROS-01
For emergencies in the US, call CHEMTREC: 800-424-9300

Section 2 - Composition, Information on Ingredients

CAS# Chemical Name Percent EINECS/ELINCS
578-95-0 9(10H)-Acridone 100 209-434-7

Section 3 - Hazards Identification

EMERGENCY OVERVIEW

Appearance: green to gold powder.
Caution! Causes respiratory tract irritation. Causes eye and skin irritation. May cause digestive tract irritation. The toxicological properties of this material have not been fully investigated.
Target Organs: None.


Potential Health Effects
Eye: Causes eye irritation.
Skin: Causes skin irritation.
Ingestion: May cause irritation of the digestive tract.
Inhalation: Dust is irritating to the respiratory tract.
Chronic: Not available.

Section 4 - First Aid Measures


Eyes: Immediately flush eyes with plenty of water for at least 15 minutes, occasionally lifting the upper and lower eyelids. Get medical aid imme diately.
Skin: Get medical aid. Immediately flush skin with plenty of water for at least 15 minutes while removing contaminated clothing and shoes.
Ingestion: Do not induce vomiting. If victim is conscious and alert, give 2-4 cupfuls of milk or water. Never give anything by mouth to an unconscious person. Get medical aid immediately.
Inhalation: Remove from exposure and move to fresh air immediately. If not breathing, give artificial respiration. If breathing is difficult, give oxygen. Get medical aid.
Notes to Physician: Treat symptomatically and supportively.

Section 5 - Fire Fighting Measures


General Information: As in any fire, wear a self-contained breathing apparatus in pressure-demand, MSHA/NIOSH (approved or equivalent), and full protective gear. During a fire, irritating and highly toxic gases may be generated by thermal decomposition or combustion.
Extinguishing Media: Use water spray, dry chemical, carbon dioxide, or alcohol-resistant foam.
Flash Point: Not available.
Autoignition Temperature: Not available.
Explosion Limits, Lower:Not available.
Upper: Not available.
NFPA Rating: Not published.

Section 6 - Accidental Release Measures


General Information: Use proper personal protective equipment as indicated in Section 8.
Spills/Leaks: Clean up spills immediately, observing precautions in the Protective Equipment section. Sweep up, then place into a suitable container for disposal. Avoid generating dusty conditions.

Section 7 - Handling and Storage


Handling: Wash thoroughly after handling. Avoid contact with skin and eyes. Avoid ingestion and inhalation. Use with adequate ventilation.
Storage: Store in a cool, dry place. Keep container closed when not in use.

Section 8 - Exposure Controls, Personal Protection


Engineering Controls: Use adequate ventilation to keep airborne concentrations low.
Exposure Limits
Chemical Name ACGIH NIOSH OSHA - Final PELs
9(10H)-Acridone none listed none listed none listed

OSHA Vacated PELs: 9(10H)-Acridone: No OSHA Vacated PELs are listed for this chemical.
Personal Protective Equipment
Eyes: Wear appropriate protective eyeglasses or chemical safety goggles as described by OSHA's eye and face protection regulations in 29 CFR 1910.133 or European Standard EN166.
Skin: Wear appropriate protective gloves to prevent skin exposure.
Clothing: Wear appropriate protective clothing to prevent skin exposure.
Respirators: Follow the OSHA respirator regulations found in 29 CFR 1910.134 or European Standard EN 149. Use a NIOSH/MSHA or European Standard EN 149 approved respirator if exposure limits are exceeded or if irritation or other symptoms are experienced.

Section 9 - Physical and Chemical Properties


Physical State: Powder
Appearance: green to gold
Odor: None reported.
pH: Not available.
Vapor Pressure: Not available.
Vapor Density: Not available.
Evaporation Rate:Not available.
Viscosity: Not available.
Boiling Point: Not available.
Freezing/Melting Point:> 300 deg C
Decomposition Temperature:Not available.
Solubility: Not available.
Specific Gravity/Density:Not available.
Molecular Formula:C13H9NO
Molecular Weight:195.22

Section 10 - Stability and Reactivity


Chemical Stability: Stable under normal temperatures and pressures.
Conditions to Avoid: Incompatible materials.
Incompatibilities with Other Materials: Strong oxidizing agents.
Hazardous Decomposition Products: Nitrogen oxides, carbon monoxide, irritating and toxic fumes and gases, carbon dioxide.
Hazardous Polymerization: Has not been reported.

Section 11 - Toxicological Information


RTECS#:      
CAS# 578-95-0: AR6976000
LD50/LC50:
Not available.

Carcinogenicity:
CAS# 578-95-0: Not listed by ACGIH, IARC, NTP, or CA Prop 65.

Epidemiology: No data available.
Teratogenicity: No data available.
Reproductive Effects: No data available.
Mutagenicity: DNA damage-Unspecified effects: lyms 10 pph
Neurotoxicity: No data available.
Other Studies:

Section 12 - Ecological Information

No information available.

Section 13 - Disposal Considerations

Chemical waste generators must determine whether a discarded chemical is classified as a hazardous waste. US EPA guidelines for the classification determination are listed in 40 CFR Parts 261.3. Additionally, waste generators must consult state and local hazardous waste regulations to ensure complete and accurate classification.
RCRA P-Series: None listed.
RCRA U-Series: None listed.

Section 14 - Transport Information

US DOT Canada TDG
Shipping Name: Not regulated as a hazardous material No information available.
Hazard Class:
UN Number:
Packing Group:

Section 15 - Regulatory Information

US FEDERAL

TSCA
     CAS# 578-95-0 is listed on the TSCA inventory.
Health & Safety Reporting List
     None of the chemicals are on the Health & Safety Reporting List.
Chemical Test Rules
     None of the chemicals in this product are under a Chemical Test Rule.
Section 12b
     None of the chemicals are listed under TSCA Section 12b.
TSCA Significant New Use Rule
     None of the chemicals in this material have a SNUR under TSCA.
CERCLA Hazardous Substances and corresponding RQs
     None of the chemicals in this material have an RQ.
SARA Section 302 Extremely Hazardous Substances
     None of the chemicals in this product have a TPQ.
Section 313      No chemicals are reportable under Section 313.
Clean Air Act:
     This material does not contain any hazardous air pollutants.
     This material does not contain any Class 1 Ozone depletors.
     This material does not contain any Class 2 Ozone depletors.
Clean Water Act:
     None of the chemicals in this product are listed as Hazardous Substances under the CWA.
     None of the chemicals in this product are listed as Priority Pollutants under the CWA.
     None of the chemicals in this product are listed as Toxic Pollutants under the CWA.
OSHA:
     None of the chemicals in this product are considered highly hazardous by OSHA.
STATE
     CAS# 578-95-0 is not present on state lists from CA, PA, MN, MA, FL, or NJ.

California Prop 65

California No Significant Risk Level: None of the chemicals in this product are listed.

European/International Regulations

European Labeling in Accordance with EC Directives
Hazard Symbols:
     XI
Risk Phrases:

     R 33 Danger of cumulative effects.
     R 36/37/38 Irritating to eyes, respiratory system and skin.

Safety Phrases:

     S 24/25 Avoid contact with skin and eyes.
     S 26 In case of contact with eyes, rinse immediately with plenty of
     water and seek medical advice.
     S 36/37/39 Wear suitable protective clothing, gloves and eye/face pr
     otection.

WGK (Water Danger/Protection)

     CAS# 578-95-0: No information available.
Canada - DSL/NDSL
     CAS# 578-95-0 is listed on Canada's NDSL List.
Canada - WHMIS
     This product has a WHMIS classification of D2B.
This product has been classified in accordance with the hazard criteria of the Controlled Products Regulations and the MSDS contains all of the information required by those regulations.
Canadian Ingredient Disclosure List

Section 16 - Additional Information

MSDS Creation Date: 3/10/1998
Revision #3 Date: 10/03/2005

The information above is believed to be accurate and represents the best information currently available to us. However, we make no warranty of merchantability or any other warranty, express or implied, with respect to such information, and we assume no liability resulting from its use. Users should make their own investigations to determine the suitability of the information for their particular purposes. In no event shall Fisher be liable for any claims, losses, or damages of any third party or for lost profits or any special, indirect, incidental, consequential or exemplary damages, howsoever arising, even if Fisher has been advised of the possibility of such damages.